Brushes, Micro Cleaning
Supplier: WORLD PRECISION INSTRUMENTS LLC
503231
77663-168EA 82.73 CAD
77663-168
Brushes, Micro Cleaning
Brushes
Non-linting, non-absorbent 2 mm diameter fiber head has a patented spherical shape that holds ¹/₈ drop of material.
  • Vial of 100