Stainless Steel Posts
Supplier: WORLD PRECISION INSTRUMENTS LLC
502191 503070 503071 503073 503075 503077
77663-066EA 43.8 CAD
77663-066 77663-086 77663-088 77663-090 77663-092 77663-094
Stainless Steel Posts
Furniture Shelves
These high quality components are made of stainless steel and polymer that resists organic solvents and corrosion. They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs.
Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable