McIndoe Dressing Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501267 501267-G 501268
77662-546EA 131.39 CAD
77662-546 77662-548 77662-550
McIndoe Dressing Forceps
Forceps
Our McIndoe Dressing Forceps have fine, narrow tips for grasping delicate tissues and for accessing deeper within an incision. Originally designed for plastic surgery, they are also popular for microdissection, and ophthalmology.
  • 15.5 cm (6") long
  • Straight
  • Serrated
  • Grooved handle for better control and lighter weight
  • Plain or Tugsten carbide inserts
The stabilizing pin and grooved handle give you a secure grip for better control of the tweezers. They come as standard or with tungsten carbide inserts. Tungsten carbide (TC) is extremely hard and durable,More Product Information