Scissors, Metzenbaum, Curved
Supplier: WORLD PRECISION INSTRUMENTS LLC
501261 501261-G 501262 501262-G
77662-522EA 328.47 CAD
77662-522 77662-524 77662-526 77662-528
Scissors, Metzenbaum, Curved
Scissors
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • 15 cm (5.9")
  • Curved, combination blades one serrated and one tungsten carbide insert
  • German
  • Blunt, narrow tips
  • Stainless steel
These are the finest scissors available. They combine the precise cutting action of the razor-sharp SuperCut blades with the durability of Tungsten Carbide. They will provide a clean, smooth cut and retain their sharpness for anMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable