Metzenbaum Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
501252 501252-G 501253 501253-G 501254 501254-G 501747 501747-G 501748 501748-G 501255
77662-504EA 99.78 CAD
77662-504 77662-506 77662-508 77662-510 77662-512 77662-514 77662-776 77662-778 77662-780 77662-782 77662-516
Metzenbaum Scissors
Scissors
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.