Ball Joint Attachment Arms for Micromanipulator
Supplier: WORLD PRECISION INSTRUMENTS LLC
500476 500475
77662-376EA 615.55 CAD
77662-376 77662-374
Ball Joint Attachment Arms for Micromanipulator
Adapters Adapters, Spherical Ground Joints
A ball joint attachment Arm for a micromanipulator gives you flexibility and precise positioning options. With a ball joint attachment you can adjust the orientation and angle of the micromanipulator's probe.
  • For use with MMJ, M325, MD4, and KITE
  • Joystick controlled manual micromanipulor MMJ
  • Micrometer slide micromanipultor M325
  • Dual tool holder micromanipulator MD4
  • Economy manual micromanipulor KITE
The ball joint allows for a 360º range of movement, including rotational movement and gives you greater maneuverability. Now you can comfortably positionMore Product Information