Universal Dipping Probe Stand
Supplier: WORLD PRECISION INSTRUMENTS LLC
42200
77662-096EA 669.08 CAD
77662-096
Universal Dipping Probe Stand
Electrode Holders
The universal dipping probe stand is designed to holds dip tips, diptip minis, spectropipetters and biosensors (macro and micro).
Delivery information: Kit includes: Small V-base, 25 cm stainless steel post and holder bracket, delrin with thumbscrew