McPherson-Vannas Spring Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
501234 501234-G 14124 14124-G
77662-450EA 693.4 CAD
77662-450 77662-452 77661-768 77661-770
McPherson-Vannas Spring Scissors
Scissors
McPherson-Vannas spring scissors were originally designed by Dr. David McPherson and Dr. Erkki Vannas in the 1970s specifically for microsurgery, which requires precision in the dissection of delicate tissues. The fine, sharp tips of these Vannas spring scissors allow for precise cutting and dissection of small tissues. For greater visibility at the tip, select the curved blades.
  • Perfect for ambidextrous use in microsurgery or fine dissection
  • Smooth cutting action with these spring scissors
  • Fine, sharp tips, either curved or straight
  • Stainless steel or
More Product Information