Webster Needle Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
14109 14109-G 501843 501929
77661-724EA 99.78 CAD
77661-724 77661-726 77662-878 77662-954
Webster Needle Holders
Forceps
Webster needle holders are surgical instrument use to hold and manipulate suturing needles. While Webster needle holders are often associated with microsurgery and microvascular procedures, they can be used in a variety of surgical contexts where precise needle handling is required.
  • Extra fine or rine straight tips
  • Serrated or smooth jaws
  • 1.2 mm tips
  • Lock
  • Surgical grade stainless steel or premium German steel
Webster needle holders have fine tips for precise control when grasping needles. The fine tips make it easier to handle and manipulate small andMore Product Information