Human/Mouse/Rat Recombinant Activin-A (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
800-01AF-100UG 800-01AF-10UG 800-01AF-1MG 800-01AF-500UG
77661-042EA 1059.44 CAD
77661-042 77661-044 77661-046 77661-048
Human/Mouse/Rat Recombinant Activin-A (Animal free) (from E. coli)
Proteins and Peptides
Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins and functions to stimulate follicle-stimulating hormone (FSH) secretion. Activins are produced in many tissue types including the skin, gonads, lungs, and pituitary gland. Activins interact with receptor type I and type II serine/threonine protein kinases, to activate SMAD signaling and regulate diverse cellular functions, such as cell proliferation, differentiation, wound healing, apoptosis, and metabolism. Activin A is a homodimer comprised of two activin βA chains. Cleavage of the N-terminal propeptide renders the Activin protein biologically active. Human Activin A shares 100% amino acid sequence identity with mouse, rat, porcine, bovine, and feline Activin A proteins.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.