VWR® Incubating Microplate Shakers
Supplier: VWR International
77623-780 77623-784 77623-788 77623-776
77623-780EA 6316.05 CAD
77623-780 77623-784 77623-788 77623-776
VWR® Incubating Microplate Shakers
Shakers Shaking Incubators
VWR® Incubating microplate shakers feature variable-speed, microprocessor control, providing reliable, uniform mixing action with accurate temperature control and are ideal for cell cultures, immunoassays, hybridization, genetics research and bacterial suspensions.
  • LCD display for temperature, speed, and time
  • Includes platform ready to secure four microplates, deep well plates, or two microtube racks
  • Available with clear lid or opaque lid for light sensitive samples
  • RS232 interface provides two-way communication for data logging and unit control
  • Durable
More Product Information