VWR® Incubating Microplate Shakers, 230 V
Supplier: VWR International
77623-782 77623-786 77623-790 77623-778
77623-782EA 6316.05 CAD
77623-782 77623-786 77623-790 77623-778
VWR® Incubating Microplate Shakers, 230 V
Shakers Shaking Incubators
VWR® Incubating Microplate Shakers feature variable-speed, microprocessor control, providing reliable, uniform mixing action with accurate temperature control you can trust and are ideal for cell cultures, immunoassays, hybridization, genetics research and bacterial suspensions.
  • 230 V models
  • LCD display for temperature, speed, and time
  • Includes platform ready to secure 4 microplates, deep well plates, or 2 microtube racks
  • Available with clear lid or opaque lid for light sensitive samples
  • RS232 interface provides two-way communication for data logging and
More Product Information