SHOWA CN751 Chemical Resistant Gloves
Supplier: Showa
CN751L-09 CN751M-08 CN751S-07 CN751XL-10 CN751XXL-11
77623-526BG 103.55 CAD
77623-526 77623-528 77623-530 77623-532 77623-534
SHOWA CN751 Chemical Resistant Gloves
Gloves
The SHOWA CN750 features a high level of chemical resistance. At 381 mm in length, the glove is designed with a grip wave pattern to ensure a secure grip during use, even in challenging conditions.
  • Oil-, chemical-resistant
  • Forearm protection
  • Impermeable and durable
  • Easy donning and doffing
  • Eco Best Technology® (EBT)
  • Unlined and wave pattern finish
The interior of the glove incorporates innovative technology. The flocking particles help to distribute moisture, making it easier to put on and take off the gloves even after extended wear. The SHOWA CN751More Product Information