VWR® 950 Benchtop pH Meter
Supplier: VWR International
77619-222 VWR-PH950A VWR-PH950AK 77619-228 77619-230 VWR-PH950DI VWR-PH950SL 77619-236 77619-238
77619-222EA 2007.95 CAD
77619-222 77619-224 77619-226 77619-228 77619-230 77619-232 77619-234 77619-236 77619-238
VWR® 950 Benchtop pH Meter
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-PH950 meter provide accurate and reliable measurements of pH, mV, ORP and temperature.
  • High resolution color touch screen (7.0")
  • 1 to 6 point calibration with standard recognition
  • Selectable pH buffer groups, including USA, NIST, DIN, GB, MERK, JIS
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 1000 sets of measurement results (GLP-compliant)
  • Automatic electrode diagnosis with pH slope and offset display
The easy to use 77619-222 benchtop meters provides
More Product Information