VWR® 900 Portable pH Meters
Supplier: SCHULER SCIENTIFIC
VWR-PH900B VWR-PH900BV VWR-PH900DI VWR-PH900SL VWR-PH900SS VWR-PH900WW VWR-PH900 VWR-PH900A VWR-PH900AK
77619-210EA 2195.35 CAD
77619-210 77619-212 77619-214 77619-216 77619-218 77619-220 77619-204 77619-206 77619-208
VWR® 900 Portable pH Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-PH900 meter provide accurate and reliable measurements of pH, mV and temperature in any environment.
  • High resolution color touch screen (4.3")
  • One to eight point calibration with standard recognition
  • Selectable pH buffer groups, including USA, NIST, DIN, GB, MERK, JIS
  • IP65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 1000 sets of measurement results
  • Rechargeable battery
  • Automatic electrode diagnosis with pH slope and offset display
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable