VWR® 750 Benchtop Conductivity Meters
Supplier: SCHULER SCIENTIFIC
VWR-EC750 VWR-EC750A VWR-EC750B VWR-EC750C
77618-972EA 1288.9 CAD
77618-972 77618-974 77618-976 77618-978
VWR® 750 Benchtop Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC750 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature within one versatile meter.
  • High resolution LCD display (5.7")
  • Three point calibration with Standard recognition
  • IP 54 rating
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 500 data sets for each parameter (GLP-compliant)
  • Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
  • Various temperature types (
More Product Information