VWR® 700 Portable Conductivity Meters
Supplier: VWR International
VWR-EC700C VWR-EC700 VWR-EC700A VWR-EC700B
77618-970EA 1428.21 CAD
77618-970 77618-964 77618-966 77618-968
VWR® 700 Portable Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC700 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature in any environment.
  • High resolution LCD display (3.5")
  • Three point calibration with Standard Recognition
  • IP65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 500 data sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable