VWR® 901 Portable Dissolved Oxygen Meter (Optical)
Supplier: SCHULER SCIENTIFIC
VWR-DO901A
77618-940EA 3328.91 CAD
77618-940
VWR® 901 Portable Dissolved Oxygen Meter (Optical)
Electrochemistry Meters Oxygen Meters
The VWR-DO901 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation with an optical DO electrode.
  • High resolution color touch display (4.3")
  • Air-saturated water or zero oxygen calibration capabilities
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 1000 sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Auto barometric pressure compensation
The easy to use VWR-DO901 portableMore Product Information