VWR® 900 Portable Dissolved Oxygen Meters
Supplier: VWR International
77618-936 77618-938
77618-936EA 1843.78 CAD
77618-936 77618-938
VWR® 900 Portable Dissolved Oxygen Meters
Electrochemistry Meters Oxygen Meters
The VWR-DO900 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
  • High resolution color touch display (4.3")
  • Air-saturated water or zero oxygen calibration capabilities
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 1000 sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Auto barometric pressure compensation
The easy to use VWR-DO900 portable meters provide accurateMore Product Information