VWR® pH Electrodes
Supplier: VWR International
77618-868 77618-890 PH43 77618-894 77618-896 PH46 77618-900 PH48 77618-904 PH50 77618-870 77618-872 77618-874 77618-876 77618-878 PH37 77618-882 PH39 77618-886 77618-888
77618-868EA 251.73 CAD
77618-868 77618-890 77618-892 77618-894 77618-896 77618-898 77618-900 77618-902 77618-904 77618-906 77618-870 77618-872 77618-874 77618-876 77618-878 77618-880 77618-882 77618-884 77618-886 77618-888
VWR® pH Electrodes
Electrodes pH Electrodes
The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.
  • Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements
  • BNC connector (BNC(Q9) or S7)
  • Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
  • Gel or liquid electrolyte fill solution
  • Includes unique serial number
  • Easily connect to VWR(R) pH meters
For additional specifications and application specific recommendations, please reference '
More Product Information