Block Rack D17
Supplier: SARSTEDT INC
93.852.171 93.852.172 93.852.173 93.852.174 93.852.175
77589-684CS 1089.33 CAD
77589-684 77589-686 77589-688 77589-690 77589-692
Block Rack D17
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 5×10 configuration
  • 17mm opening diameter
  • Can be autoclaved at 121 °C
  • Available with or without handle
  • Range of colors for each area of the laboratory
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
The Block Rack D17 is designed for space-saving sample storage. This rack features 50 apertures for S-More Product Information