VWR® VWB2 Water Baths, 120 V
Supplier: VWR International
77587-150 77587-152 77587-154 77587-156 77587-158
77587-150EA 1611.6 CAD
77587-150 77587-152 77587-154 77587-156 77587-158
VWR® VWB2 Water Baths, 120 V
Baths Water Baths, Unstirred
Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 L models feature a drain tap for convenient emptying and are suitable for use with water or alternatively with heat transfer beads. Baths with a capacity of 5 L or more have a heat transfer bead mode which is activated through the main menu. A drain stopper (plug) is supplied as on optional accessory to ensure heat transfer beads do not get stuck in the drain outlet. Note: Baths are classified Safety Class 1, allowing unattended operation when used with non
More Product Information