Single Edge Blades, Unbacked, Unwrapped
Supplier: Accutec Blades
94-0474-0000
77587-122CS 667.76 CAD
77587-122
Single Edge Blades, Unbacked, Unwrapped
Razors and Blades
AccuTec Unbacked single edge blades feature 3 facet edges for sharpness.
  • Unbacked blades
  • Unwrapped blades for high volume use
  • 3 facet edge for increased sharpness
  • High quality stainless steel blade
  • Made in USA
Packaging: Each box contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.