SteriPlast® Detectable Spoons, Curved, Long Handle, Blue
Supplier: BURKLE INC
5378-5012
77581-260PK 73.52 CAD
77581-260
SteriPlast® Detectable Spoons, Curved, Long Handle, Blue
Spoons
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
  • PS detectable
  • Individually packaged and sterilized, for single use
  • With curved spoon form, handle length 300 mm
  • Can be detected by color scanners, metal
More Product Information