NANOCOLOR® Photometric Tube Tests, Iron
Supplier: CTL SCIENTIFIC SUPPLY CORP
985037
77578-508EA 108.04 CAD
77578-508
NANOCOLOR® Photometric Tube Tests, Iron
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of iron in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.