Mini Hotplate, Slimline
Supplier: StatLab
SI-SDH-MINI
77578-308EA 1133.38 CAD
77578-308
Mini Hotplate, Slimline
Heaters Hot Plates
The StatLab slimline mini hotplate has a very small footprint, making it ideal for use at the microtome. It is primarily used to remove stubborn wrinkles from tissue sections, and re-orientating embedded tissue.
  • Ultra-fast heating system for rapid warm up
  • Small footprint
  • Anti-microbial powder coat finish
  • Features a matte black heated surface to provide a visual contrast for tissue sections
It may also be used in microbiology for heat fixing bacterial smears.
The body of the mini hotplate is coated with an antimicrobial finish which inhibits growthMore Product Information