Pure uTIP™ Pipette Tips for Universal Pipettors
Supplier: Biotix
63305403 63305404 63305405 63305406 63305407 63305408 63305409 63305410 63305411 63305412 63305413 63305414 63305415 63305416 63305417 63305418 63305419 63305420 63305421 63305422 63305423 63305424 63305425 63305426 63305427 63305428 63305429 63305430 63305431 63305432
77575-864CS 603.69 CAD
77575-864 77575-866 77575-868 77575-870 77575-872 77575-874 77575-876 77575-878 77575-880 77575-882 77575-884 77575-886 77575-888 77575-890 77575-892 77575-894 77575-896 77575-898 77575-900 77575-902 77575-904 77575-906 77575-908 77575-910 77575-912 77575-914 77575-916 77575-918 77575-920 77575-922
Pure uTIP™ Pipette Tips for Universal Pipettors
Pipette Tips
Enhance the performance and accuracy of experiments and improve global health with Pure uTIPs.
  • Compatible with universal pipettes
Pure utips are made with pure resin and patented technologies to improve accuracy and precision. Flexfit™ lowers necessary insertion and ejection force by ≈70% to make a secure seal, Blade™ lowers the chance of hanging droplet formation, and each tip is free of RNase, DNase, DNA, endotoxins, and nucleic acids. Same great tip and better for the environment.
Pure resin is made of 100% virgin polypropylene. No additives areMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable