VWR® Reagent Reservoirs
Supplier: VWR International
77575-238 77575-242 77575-246 77575-248 77575-250 77575-252 77575-256 77575-278 77575-280 77575-282 77575-284 77575-286 77575-260 77575-262 77575-264 77575-266 77575-268 77575-270 77575-272 77575-274 77575-276
77575-238CS 176.49 CAD
77575-238 77575-242 77575-246 77575-248 77575-250 77575-252 77575-256 77575-278 77575-280 77575-282 77575-284 77575-286 77575-260 77575-262 77575-264 77575-266 77575-268 77575-270 77575-272 77575-274 77575-276
VWR® Reagent Reservoirs
Reagent Reservoirs
Reagent reservoirs are designed for efficient liquid transfer, making them an ideal choice for repetitive pipetting tasks. These reagent reservoirs for multichannel pipettes provide a trough wide enough to fit 8 and 12 channel pipettes.
  • Trough design with graduations on the inside wall and pour-off spouts on four corners
  • Slightly tilted inner surface helps reduce residue
  • Non sterile or sterilized options available, achieving SAL 10⁻⁶ (ISO 11137)
  • Reservoirs are lot-certified, non-pyrogenic, DNase- and RNase-free
VWR® reagent reservoirs are an excellentMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M