Cut Resistant Gloves, A4 Cut Level
Supplier: Showa
XC810L-08 XC810M-07 XC810S-06 XC810XL-09 XC810XXL-10
77575-120CS 1040.76 CAD
77575-120 77575-160 77575-122 77575-136 77575-138
Cut Resistant Gloves, A4 Cut Level
Gloves
ANSI cut level A4 the new XC810 combines an excellent grip, superb tactility, 18-gauge HPPE A4 cut-resistance and touchscreen compatibility for a multitude of industrial applications.
  • HPPE reinforced
  • Ergonomic
  • Microporous nitrile coating
  • Embossed grip
  • Cut-resistant/lightweight
  • Flexible/breathable
  • Increased dexterity
  • Seamless knit/form-fitting
  • Abrasion-resistant
  • Touch screen compatible
  • Natural rubber latex free
Application in light assembly of oil-coated pieces, maintenance, mechanical and engineering, repair glass and windows, component assembly.