VWR® Premium CO₂ Incubators
Supplier: BINDER INC PL
9040-0195 77574-814 9040-1195 9040-1196
77574-812EA 11707.8 CAD
77574-812 77574-814 77574-816 77574-818
VWR® Premium CO₂ Incubators
Incubators CO₂ Incubators
The VWR® Premium CO₂ incubators are suitable for routine cell cultivation applications in the field of cell production, particularly in academic settings. They are contamination proof due to the 180 °C hot air sterilization cycle and feature highly pH stable conditions thanks to drift-free infrared CO₂ measurements conducted in real time.
  • Reliable anti-contamination concept with 180 °C hot air sterilization
  • Humidification system with condensation protection
  • Internal data logger, measured values can be downloaded in open format via USB
  • Tightly closing inner
More Product Information