Hazardous Waste Bag

Supplier: GRAINGER
3WNA3
77574-720PK 80.94 CAD
77574-720
Hazardous Waste Bag
Bags Waste Disposal Bags
  • Bottom seal type: Flat
  • Width: 12"
  • Max. load: 8 lbs.
  • Max. temperature: 160 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR