PIG® Dryer Safe Reusable Water Absorbent Sock
Supplier: New Pig
WTR019-GY
77574-712BX 206.98 CAD
77574-712
PIG® Dryer Safe Reusable Water Absorbent Sock
Spill Control Materials
Launderable sock blends into your environment while absorbing condensation and small water leaks, sock can be machine-washed and dried up to 50 times.
  • For use with water
  • Reusable
  • Machine-washable and dryer-safe
  • Available in aesthetically appealing tan and gray color options to blend into surroundings
Place this sock on windowsills or around coolers, freezers or air conditioners or in other damp areas to absorb condensation and small leaks. Soak up water, then toss in the washing machine and dryer to reuse. Ideal for use in schools, hospitals,More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable