Biosafety Cabinets, Cell Logic Class II, Type B2
Supplier: Labconco
323481002 323481012 323481102 323481112 323681002 323681012 323681102 323681112 343481002 343481012 343481102 343481112 343681002 343681012 333481002 333481012 333481102 333481112 333681002 333681012 333681102 333681112 343681102 343681112
77574-190EA 39697.5 CAD
77574-190 77574-398 77574-192 77574-400 77574-294 77574-502 77574-296 77574-504 77574-198 77574-406 77574-200 77574-408 77574-302 77574-510 77574-194 77574-402 77574-196 77574-404 77574-298 77574-506 77574-300 77574-508 77574-304 77574-512
Biosafety Cabinets, Cell Logic Class II, Type B2
Fume Hoods and Enclosures Biological Safety Cabinets
Cell logic Class II, Type B2 Biosafety Cabinets (BSCs) offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. Type B2 BSCs, sometimes referred to as 100% exhaust or total exhaust BSCs require a dedicated exhaust system and remote blower for each cabinet. Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable