Biosafety Cabinets, Cell Logic Class II, Type A2
Supplier: Labconco
322491002 322491012 322491102 322491112 322591002 322691102 322691112 332491102 332491112 332591002 332591012 332591102 332591112 332691002 332691012 332691102 332691112 342691002 342691012 342691102 342691112 322391002 322391012 322391102 322391112 332391002 332391012 332391102 332391112 332491002 332491012 342391002 342391012 322591012 322591102 322591112 322691002 322691012 342391102 342391112 342491002 342491012 342491102 342491112 342591002 342591012 342591102 342591112
77574-178EA 28771.8 CAD
77574-178 77574-386 77574-180 77574-388 77574-240 77574-284 77574-492 77574-184 77574-392 77574-244 77574-452 77574-246 77574-454 77574-286 77574-494 77574-288 77574-496 77574-292 77574-498 77574-290 77574-500 77574-136 77574-344 77574-138 77574-346 77574-140 77574-348 77574-142 77574-350 77574-182 77574-390 77574-144 77574-352 77574-448 77574-242 77574-450 77574-282 77574-490 77574-146 77574-354 77574-186 77574-394 77574-188 77574-396 77574-248 77574-456 77574-250 77574-458
Biosafety Cabinets, Cell Logic Class II, Type A2
Fume Hoods and Enclosures Biological Safety Cabinets
Cell logic Class II, Type A2 Biosafety Cabinets (BSCs) offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. Cell logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.
  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted)
More Product Information