SoLo™ Hydraulic Base Stands
Supplier: Labconco
5185310 5185400 5185410 5185500 5185510 5185600 5185610 5185300
77574-384EA 11706.35 CAD
77574-384 77574-238 77574-446 77574-280 77574-488 77574-342 77574-550 77574-176
SoLo™ Hydraulic Base Stands
Fume Hoods and Enclosures PCR Workstation Enclosures
SoLo™ hydraulic base stand for Labconco Class II Biosafety cabinets, optic white.
  • For all optic white Labconco Class II BSC models
  • Hydraulic lift provides flexible height adjustment and permits passage through standard doorways as low as 191.7 cm (75.5") high for A2 cabinets
  • Infinitely adjustable height provides work surface heights from 43.2 cm (17.0") to 94.0 cm (37")
  • Supports loads up to 1,000 lbs.
  • 12.7 cm (5") dia, toe-locking, glass-filled nylon casters; resistant to most chemicals, easy-clean and maintenance-free
  • Durable epoxy-coated steel
More Product Information