PuriCare Procedure Stations
Supplier: Labconco
312320002 312320012 312321002 312321012 312420002 312420012 312421002 312421012 312520002 312520012 312521002 312521012 312620002 312620012 312621002 312621012
77574-172EA 20903.4 CAD
77574-172 77574-380 77574-174 77574-382 77574-234 77574-442 77574-236 77574-444 77574-276 77574-484 77574-278 77574-486 77574-338 77574-546 77574-340 77574-548
PuriCare Procedure Stations
Fume Hoods and Enclosures Biological Safety Cabinets
PuriCare procedure stations offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. These Class II, Type A2 biosafety cabinets are ideal for small animal research activities including surgical procedures, cage changing, weighing, and anesthesia delivery. All models ensure the highest standard of safety and performance and are listed to NSF/ANSI 49.
  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable