Biosafety Cabinets, Logic Class II, Type A2
Supplier: Labconco
302310002 302310012 302310102 302310112 302311002 302311012 302421002 302421012 302421102 302421112 302480002 302480012 302480102 302611002 302611012 302611102 302611112 302620002 302620012 302620102 302620112 302621002 302621012 302410102 302410112 302411002 302411012 302411102 302411112 302420002 302420012 302420102 302380002 302380012 302380102 302380112 302381002 302381012 302381102 302381112 302410002 302410012 302481102 302481112 302510002 302510012 302510102 302510112 302511002 302511012 302511102 302511112 302520002 302520012 302520102 302520112 302521002 302521012 302521102 302521112 302580002 302580012 302311102 302311112 302320002 302320012 302320102 302320112 302321002 302321012 302321102 302321112 302420112 302621102 302621112 302680002 302680012 302680102 302680112 302681002 302681012 302681102 302681112 302480112 302481002 302481012 302580102 302580112 302581002 302581012 302581102 302581112 302610002 302610012 302610102 302610112
77574-150EA 20800.7 CAD
77574-150 77574-358 77574-148 77574-356 77574-152 77574-360 77574-214 77574-422 77574-216 77574-424 77574-220 77574-428 77574-218 77574-310 77574-518 77574-312 77574-520 77574-316 77574-524 77574-314 77574-522 77574-318 77574-528 77574-202 77574-410 77574-206 77574-414 77574-208 77574-416 77574-212 77574-420 77574-210 77574-166 77574-374 77574-164 77574-372 77574-168 77574-376 77574-170 77574-378 77574-204 77574-412 77574-224 77574-432 77574-254 77574-462 77574-252 77574-460 77574-256 77574-464 77574-258 77574-466 77574-262 77574-470 77574-260 77574-468 77574-264 77574-472 77574-266 77574-474 77574-270 77574-478 77574-154 77574-362 77574-158 77574-366 77574-156 77574-364 77574-160 77574-368 77574-162 77574-370 77574-418 77574-320 77574-526 77574-324 77574-532 77574-322 77574-530 77574-326 77574-534 77574-328 77574-536 77574-426 77574-222 77574-430 77574-268 77574-476 77574-272 77574-480 77574-274 77574-482 77574-308 77574-516 77574-306 77574-514
Biosafety Cabinets, Logic Class II, Type A2
Fume Hoods and Enclosures Biological Safety Cabinets
Logic Class II, Type A2 Biosafety Cabinets (BSCs) offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1 to 4 containment. These cabinets are ideal for applications involving biological hazards, genetic material, antineoplastic drugs and other hazardous airborne particulates. All models ensure the highest standard of safety and performance.
  • Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5" line-of-sight touchscreen display
More Product Information