SymLine HPLC Waste Bundles, V3.0
Supplier: SCAT AMERICAS, INC.
75312 75300 75301 75302 75310 75311
77553-688EA 5819.3 CAD
77553-688 77553-678 77553-680 77553-682 77553-684 77553-686
SymLine HPLC Waste Bundles, V3.0
Chromatography Supplies Chromatography Spare Parts
SymLine FLEX bundle.
Certifications: UN-Y approved
Delivery information: Supplied with 1×Basic 32 pipe, PE-HD electrostatic conductive, 1×GL 45 (m), 1×GL 25 (m), pipe connection (w), length: 310 mm, tensile strength (Rm) = 400 N, tightening torque = 4 Nm; 1×SymLine FlexTube, Length 1000 mm - flexible adaptable; 1×Shut-off, angled, 1×Exhaust filter M, V3.0; 1×Offset adapter 90° for exhaust filter; 1×Safety waste cap, S60/61, GL25 (m), connector for exhaust filter, PE-HD electrostatic conductive; 2×10 L / 2.64 gallon multilayer HDPE container with handle, natural colour. UN-Y approved,More Product Information