Homogenizer, Bead Ruptor 24
Supplier: OMNI INTERNATIONAL
19-070
77547-556EA 13452.12 CAD
77547-556
Homogenizer, Bead Ruptor 24
Homogenizers Benchtop Homogenizers
The bead ruptor 24 homogenizer is ideal for demanding laboratories seeking unmatched reliability, power and efficiency in grinding, lysing and homogenization of even the toughest samples.
  • Fixed 2 ml tube carriage - process 24×2 ml tubes
  • Convenient front loading design
  • Programmable memory settings
  • Optimized tube motion ensures the highest performance
  • Ideal for: DNA/RNA extraction, tissue homogenization, enzyme isolation and protein purificaion
The bead ruptor 24 bead mill homogenizer is designed with a unique carriage motion to ensure that the intra-More Product Information