Recombinant Bacteriophage T4 UvsX DNA Recombinase (Glycerol Free or Lyophilized) (from E. coli), His Tag
Supplier: G-Biosciences
786-2131 786-2132 786-2133 786-2134 786-2135 786-2136
77540-320EA 525.08 CAD
77540-320 77540-322 77540-324 77540-326 77540-328 77540-330
Recombinant Bacteriophage T4 UvsX DNA Recombinase (Glycerol Free or Lyophilized) (from E. coli), His Tag
Enzymes
T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.
  • It is used for Recombinase Polymerase Amplification (RPA) assay
Rapid diagnostic assay.