Gram's iodine solution Avanced

Supplier: G-Biosciences

786-2097 786-2098
77540-252KT 175.76 CAD
77540-252 77540-254
Gram's iodine solution Avanced
Gram's iodine solution

Gram Iodine Advanced Solution acts as a mordant .


  • An aqueous solution of Iodine


It is used in Gram staining to differentiate gram-positive from gram-negative bacteria

Formula: I₃K
MDL Number: MFCD00134128
CAS Number: 12298-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR