Crystal violet solution for Gram staining

786-2079 786-2080 786-2081 786-2082
77540-216EA 468.49 CAD
77540-216 77540-218 77540-220 77540-222
Crystal violet solution for Gram staining
Crystal Violet

A primary stain in the advanced Gram stain procedure.


  • An aqueous solution of crystal violet


Quick staining of Gram-positive and Gram-negative bacteria

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR