Acridine orange (hemizinc salt) solution

Supplier: G-Biosciences

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

786-2074
77540-206EA 48.74 CAD
77540-206
Acridine orange (hemizinc salt) solution
Acridine orange (hemizinc salt)

Acridine Orange is a cell-permeant vital dye that binds to nucleic acids.


It is used in the fluorescent microscopic detection of microorganisms from clinical samples.


Binding to dsDNA causes acridine orange to fluoresce green and binding to ssDNA or RNA causes it to fluoresce red.

Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
MDL Number: MFCD00081043
CAS Number: 10127-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR