Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
FLO-MIN106D FLO-MIN114 FLO-MIN114.8 FLO-MIN004RAA FLO-MIN004RAA.8 FLO-MIN004RAA.8.48 FLO-MIN004RAA.8.96 FLO-MIN114.8.48 FLO-MIN114.8.96
76487-106EA 1106 CAD
76487-106 76519-502 76521-802 76521-804 76595-418 76595-414 77308-570 77532-276 77532-278 77532-280 77532-282 77532-284
Flow Cells for MinION™ and GridION™ Devices
Flow Cells
Nanopore sequencing flow cells for use with MinION™ and GridION™ devices.
Flow cells contain the proprietary sensor array, Application-Specific Integrated Circuit (ASIC), and nanopores for DNA/RNA sequencing on MinION™ and GridION™ devices.
R10.4.1 flow cells contain a double reader-head and are suitable for experiments where high consensus accuracy is required. Compatible with Kit 14 chemistry, R10.4.1 flow cells generate data at a modal accuracy above 99%. R9.4.1 flow cells are compatible with the previous generation of nanopore sequencing chemistries. These flow cells and associatedMore Product Information
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.