Spunbond Coveralls
Supplier: O&M HALYARD CANADA, INC CA
85631 85641 85651 85661
77531-672CS 93.6 CAD
77531-672 77531-674 77531-676 77531-678
Spunbond Coveralls
Disposable Garments
HALYARD* Spunbond coveralls are manufactured with lightweight polypropylene spunbond fabric for breathability and wearer's comfort. These coveralls are appropriate for situations in which light fluid contact or contact isolation is anticipated.
  • For low exposure to fluids or chemicals
  • Spunbond fabric
  • Knit cuffs
  • Open legs
  • Full zip front
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Fabric Spunbond Polypropylene
Closure type Zip closure
Pockets Without pockets
Gender
Sleeve or leg length Long
Seam type Stitched
Garment application Chemical protection