Protective Coveralls
Supplier: O&M HALYARD CANADA, INC CA
75641 75651 75631
77531-668CS 279 CAD
77531-668 77531-670 77531-666
Protective Coveralls
Disposable Garments
HALYARD* Protective coveralls are appropriate for situations in which light fluid contact or contact isolation is anticipated. Features include a soft breathable 3-layer SMS fabric plus tailored styling and fit to enhance worker comfort.
  • Raglan sleeve
  • Protective 3-layer SMS fabric
  • For low exposure to fluids or chemicals
  • Traditional collar
  • Single use
  • Recommended areas for product use: Central services construction cleaning and maintenance