PureClean™ Tyvek® 1421B Autoclavable Stopper Bowl Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BCTK50 BCTK52 BCTK52-2PLY12 BCTK54 BCTK56 BCTK60 BCTK64 BCTK72 BCTK80 BCTK90 BCTK120 BCTK144 BCTK20 BCTK24 BCTK26 BCTK30 BCTK32 BCTK32-2PLY8 BCTK34 BCTK36 BCTK36-2PLY8 BCTK38 BCTK38-2PLY8 BCTK40 BCTK42 BCTK42-2PLY10 BCTK42-2PLY12 BCTK44 BCTK46 BCTK48
77531-140CS 4021.31 CAD
77531-140 77531-158 77531-148 77531-118 77531-142 77531-124 77531-130 77531-128 77531-122 77531-170 77531-166 77531-136 77531-132 77531-146 77531-120 77531-168 77531-162 77531-152 77531-172 77531-134 77531-144 77531-116 77531-174 77531-154 77531-150 77531-160 77531-138 77531-126 77531-164 77531-156
PureClean™ Tyvek® 1421B Autoclavable Stopper Bowl Covers
Covers
PureClean™ autoclave stopper bowl covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® on both sides for maximum breathability during sterilization
  • Eliminate the chance of residue deposits on equipment post sterilization
  • Manufactured and packaged in a
More Product Information