CIP 150® Alkaline Process and Research Cleaners
Supplier: Steris
1D1505 1D1502 1D15D3
CA14004-232EA 405.17 CAD
CA14004-232 77531-106 77531-098
CIP 150® Alkaline Process and Research Cleaners
Cleaning Agents Detergents and Surface Cleansers
CIP 150® Alkaline detergent is a proprietary blend of potassium hydroxide, sodium hypochlorite (oxidizing agent), surfactants and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.
  • Flexibility - Effective cleaning against a wide variety of organic and proteinaceous process residues in clean-in-place (CIP), clean-out-of-place (COP)
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable