SatPax® 670 AS (Aerospace) Pre-Wetted Non-Woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Supplier: Berkshire
SPX67000112AS
77528-882CS 538.8 CAD
77528-882
SatPax® 670 AS (Aerospace) Pre-Wetted Non-Woven Cleanroom Wiper, 55% Cellulose/45% Polyester
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® 670 AS for Aerospace combines Durx® 670 non-woven wipers with 70% IPA/30% DI water to create a pre-wetted wiper that is effective and easy to use.
  • Low extractables and fiber and particle counts
  • Smooth and durable with good wet strength
  • Reduces alcohol usage and preparation/handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency and cleaning protocol consistency
  • Pre-wetted with consistent 70% IPA/30% DI water to a saturation level of 45%
  • Re-sealable solvent-resistant packaging
  • C-fold packaging configuration for single withdrawal