Human FA (Folic Acid) ELISA Kit

Supplier: AFG Bioscience

EK248344
77527-104EA 609.41 CAD
77527-104
Human FA (Folic Acid) ELISA Kit
Assays ELISAs

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein FA (Folic Acid)
Primary antibody reactivity
Assay Type Competitive
Host Human
Sensitivity 14.7 pg/ml
Detection range 156.25 - 10000 pg/ml
Sample type Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
Time to Results 2 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Hematology, reproductive science, nutrition metabolism

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR